Hydrochloric acid 3 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
4.80680.9025
: Merck
:
- Pack type:Plastic bottle
- Pk:25 L
- Seq:5327
:
:1.06 g/cm³ (20 °C)
:231-595-7
:8,II
:36.46 g/mol
:1789
:GHS05
:GHS07
:Ambient
:7647-01-0
:HCl
:MFCD00011324