
Hydrochloric acid 0.1 mol/l (0.1 N) in ethanol
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:3,II
:36.46 g/mol
EINECS:231-595-7
UN:1993
:GHS02
Storage Temperature:Ambient
Frequently Bought Together