Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride 0.1 M HCl (0.1N) in 2-propanol volumetric solution, for total base number, according to DIN 51558-1, Fluka™
Irritant
Corrosive
Flammable
Hydrogen chloride 0.1 M HCl (0.1N) in 2-propanol volumetric solution, for total base number, according to DIN 51558-1, Fluka™
Catalog # 35009-1L
CAS Number:  
Irritant
Corrosive
Flammable
Hydrogen chloride 0.1 M HCl (0.1N) in 2-propanol volumetric solution, for total base number, according to DIN 51558-1, Fluka™
Catalog # 35009-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    0.099 - 0.101 mol/l
  • Water (Karl Fischer):
    Max. 1 %
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :36.46 g/mol
Boiling Pt:82 °C
Density:0.788 g/cm³
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324