(±)-α-Methoxyphenylacetic acid, Sigma-Aldrich®
Catalog # SIAL248975-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:166.18 g/mol
Boiling Pt:165 °C (24 hPa)
Melting Pt:69…71 °C
:M-Clarity™ quality level = MQ200
EINECS:230-300-9
CAS Number:7021-09-2
Formula:C₉H₁₀O₃
MDL Number:MFCD00064217