Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Bovine native beta-Casein (from Milk)
Bovine native beta-Casein (from Milk)
:
Merck
Bovine native beta-Casein (from Milk)
:
Merck
CAS No.:
9000-71-9
Protein/peptide type:
Native
Source:
Milk
Species:
Bovine
MDL No.:
MFCD00081481
Protein/peptide name:
beta-Casein