



Borane dimethyl sulphide complex 2 mol/l in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13292-87-0
:
Formula:C₂H₉BS
MDL Number:MFCD00013189
:75.97 g/mol
Flash Pt:–17 °C
Storage Temperature:Refrigerator
EINECS:236-313-6
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
:GHS08
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:1.8 to 2.2 M
- Loss on drying:4 % to 9 % (1 g, 105°C) (vacuum)
- HPLC:≥97.5 %
- Specific optical rotation:+36° to +38° (20°C, 589 nm) (1 % in water)