



Borane dimethyl sulphide complex 1 mol/l in dichloromethane, AcroSeal®
Catalog # 185611000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.95 to 1.05 M
- Water:≤3 %
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:75.97 g/mol
Flash Pt:18 °C
EINECS:236-313-6
UN:3399
:GHS02
:GHS08
:GHS05
:GHS07
Storage Temperature:Refrigerator
CAS Number:13292-87-0
Formula:C₂H₉BS
MDL Number:MFCD00013189