



Borane dimethyl sulphide complex ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13292-87-0
:
Formula:C₂H₉BS
MDL Number:MFCD00013189
:75.97 g/mol
Boiling Pt:∼44 °C (dec.)
Melting Pt:–38 °C
Density:0.801
Flash Pt:–1 °C (30 °F)
EINECS:236-313-6
UN:2924
ADR:3,II
:GHS02
:GHS06
:GHS05
:GHS07