Heavy-duty Tracks
Supplier: WALDNER LABOREINRICHTUNG
Certificates
About this item
These tracks are suitable for holding stackable and non-stackable measuring instruments/measuring device towers.
- Mobile multi-level storage area
- With flexible, horizontally freely configurable work surfaces
- Suitable for heavy equipment
- Shelf made of coated OSB board
- Steel profile 70×40 mm
These tracks feature fixed lower shelf, two height adjustable shelves, steel frame, four castors, of which two are lockable (roller and steering axle lockable).