Order Entry
Germany
ContactUsLinkComponent
Microtitre Plates, FlowerPlate, pH/DO 2 (LG1/RF)
Microtitre Plates, FlowerPlate, pH/DO 2 (LG1/RF)
Catalog # BECKM2P-MTP-48BOH2
Supplier:  Beckman Coulter
Microtitre Plates, FlowerPlate, pH/DO 2 (LG1/RF)
Catalog # BECKM2P-MTP-48BOH2
Supplier:  Beckman Coulter

Specifications

  • No. of wells:
    48
  • Description:
    FlowerPlate microtitre plate, pH/DO type 2 (LG1/RF)
  • Pk:
    1

Specifications

About this item

The FlowerPlate microtiter plate is a 48-well MTP in standard ANSI/SLAS (SBS) format. Its flower-shaped wells resemble baffles in shake flasks and facilitate high gas transfer into the liquid phase.

This FlowerPlate MTP has two optical sensors (optodes) per cultivation well, i.e. the LG1 optode for measuring the pH value and the RF optode for measuring the DO.

The following bioprocess parameters can be read out online by the BioLector microbioreactor:
• Biomass
• pH
• Dissolved oxygen (DO)
• Fluorescence

This MTP is compatible with: BioLector II, BioLector Pro and BioLector XT microbioreactors.