Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid 96%
2-Chloro-5-pyridineboronic acid 96%
2-Chloro-5-pyridineboronic acid 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2-Chloro-5-pyridineboronic acid 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
444120-91-6
:
Formula:
C₅H₅BClNO₂
MDL Number:
MFCD03094998
:
157.36 g/mol
Melting Pt:
154…157 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL20388
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation