Cholesterol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-88-5
:
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646
:386.66 g/mol
Merck Index:13,02221
Boiling Pt:360 °C (1013 hPa)
Melting Pt:147…150 °C
Density:1.07 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
EINECS:200-353-2