Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4-methylbenzonitrile
2-Amino-4-methylbenzonitrile, Sigma-Aldrich®
2-Amino-4-methylbenzonitrile, Sigma-Aldrich®
2-Amino-4-methylbenzonitrile, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-Amino-4-methylbenzonitrile, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
26830-96-6
:
Formula:
C₈H₈N₂
MDL Number:
MFCD00173706
:
132.17
ADR:
6.1,III
:
M-Clarity™ quality level = MQ100
EINECS:
248-020-0
UN:
3439