Order Entry
Germany
Orders LinkContactUsLinkComponent
Propylene oxide

Propylene oxide

1
Catalog # POLS00236-1
Flammable
Health
Irritant
Marketsource
Supplier:   POLYSCIENCES
Propylene oxide
Catalog # POLS00236-1
Supplier:  POLYSCIENCES
Marketsource
CAS Number:  75-56-9
Propylene oxide
Propylene oxide

1

CAS Number:  75-56-9
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Synonyms:  (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
  • CAS Number:75-56-9
  • Formula:C₃H₆O
  • MDL Number:MFCD00005126
  • Molecular Weight:58.08 g/mol
  • Merck Index:12,08041
  • Boiling Pt:34.3 °C (1013 hPa)
  • Melting Pt:–112 °C
  • Density:0.83 g/cm³ (20 °C)
  • Flash Pt:–37 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-879-2
  • UN:1280
  • ADR:3,I
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.