Order Entry
Germany
ContactUsLinkComponent
L-Phenylalanine
L-Phenylalanine
Catalog # MOLEM12455823
Supplier:  Molekula
CAS Number:  
L-Phenylalanine
Catalog # MOLEM12455823
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :165.19 g/mol
Merck Index:14,07271
Boiling Pt:313 °C (1013 hPa)
Melting Pt:173 °C
Density:0.754 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-568-1
CAS Number:63-91-2
Formula:C₉H₁₁NO₂
MDL Number:MFCD00064227