L-Phenylalanine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:63-91-2
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00064227
:165.19 g/mol
Merck Index:14,07271
Boiling Pt:313 °C (1013 hPa)
Melting Pt:173 °C
Density:0.754 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:200-568-1