XRF Sample Cup, Double Open-End with Baffle Plus™ Cap
XRF sample cup, double open ends with Baffle Plus™ cap, ext.Ø 31 mm, int.Ø 25 mm, height 29 mm, Vol. 6,8 ml
Catalog # PROOVHG-CUP69-31S
Supplier: LGC Standards PROMOCHEM
XRF Sample Cup, Double Open-End with Baffle Plus™ Cap
Catalog # PROOVHG-CUP69-31S
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Double open-end XRF sample cups with Baffle Plus™ cap consists of four parts.
Specifications
- Description:XRF sample cup, double open ends with Baffle Plus™ cap, ext.Ø 31 mm, int.Ø 25 mm, height 29 mm, Vol. 6,8 ml