Hei-VAP Industrial - All-round Package Rotary Evaporators
Catalog # HEID518-41100-00
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Description:Hei-VAP industrial - all-round package
- Pk:1
Specifications
About this item
Hei-VAP Industrial - All-round Package.
- This packages includes:
- Hei-VAP industrial model with glassware R - Recommended for all standard distillations
- Base cart
- Tubing set with 16 meter piece incl. hose clamps
- Vacuum pump Hei-VAC valve industrial
- Chiller Hei-CHILL 3000
- 40 L - Kryo 30 thermal fluid