Vacuum desiccators cabinets, SICCO
Supplier: Bohlender
Certificates
About this item
These transparent acrylic cabinets have solid 25 mm thick walls, a circular silicone seal and a vacuum gauge (0 to 760 mm of mercury).
- Vacuum up to 0,0001 Torr
- Temperature resistance from −20 to +70 °C
- Non slip rubber feet
- Two needle valves with Ext.Ø 9 mm
Suitable for storing sensitive products in a vacuum. The vacuum inside the desiccator ensures rapid drying of stored products. These systems allow storage in an oxygen- and carbon dioxide- free environment and are, for example, suitable for storage of semiconductor samples.
Delivery: Supplied with two or four removable shelves. Capacity to accommodate maximum of three shelves (16 L cabinet) or eight shelves (22 and 29 L cabinets).