Orders LinkContactUsLinkComponent
Mobile Tool Cabinets
Mobile Tool Cabinets
Supplier:   TEKNOMEK
Mobile Tool Cabinets
Supplier:  TEKNOMEK
Certificates
Mobile Tool Cabinets
2 Options Available Below
Product Details & Documents
This heavy duty mobile stainless steel tool cabinet with 3, 4 or 11 drawers has a brushed dull polish finish is excellent for tool storage within the workplace. With a floor clearance of 100 mm, there is plenty of room for a mop or broom for easy cleaning below the tool cabinet.
  • Easy tool organisation
  • Fully welded 304L-grade stainless steel
  • Brushed dull polish finish
  • Stainless steel castors (2 fixed and 2 locking swivel)
  • Lockable drawers
Designed with lockable drawers of varying sizes this storage cabinet provides easy and safe compartmentalisation of different types of tools for efficiency and safety.
Product Family Options
Product Information
  • Material
    W×D×H
    Availability
    Price
  • TEKMSHCCCATO4D1050
    Mobile tool cabinet with 4 drawers
    Mobile tool cabinet with 4 drawers
    Stainless steel 304L
    1000×500×840 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • TEKMSHCCCATO111050
    Mobile tool cabinet with 11 drawers
    Mobile tool cabinet with 11 drawers
    Stainless steel 304L
    1050×500×1040 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST