Order Entry
Germany
Orders LinkContactUsLinkComponent
Zinc Standard Solution

Zinc Standard Solution

Zinc standard solution, 1000 mg/L, 500 mL
Catalog # 713-4095
Supplier:   WTW
Zinc Standard Solution
Catalog # 713-4095
Supplier:  WTW
CAS Number:  
Zinc Standard Solution
Zinc Standard Solution

Zinc standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
57,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    500 mL
  • Description:
    Zinc standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.