Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
C16 dihydro Ceramide-d9 (d18:0/16:0-d9)
C16 dihydro Ceramide-d9 (d18:0/16:0-d9), solid
C16 dihydro Ceramide-d9 (d18:0/16:0-d9), solid
C16 dihydro Ceramide-d9 (d18:0/16:0-d9), solid
Catalog # CAYM39389-1
Supplier:
Cayman Chemical
CAS Number:
Expand image
C16 dihydro Ceramide-d9 (d18:0/16:0-d9), solid
Catalog # CAYM39389-1
Supplier:
Cayman Chemical
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
548.97 g/mol
CAS Number:
2260669-52-9
Formula:
C₃₄H₆₀D₉NO₃
Product Details