Bacillol® Wipes
Stand-up pouches (90 wipes), White
Supplier: BODE CHEMIE
Jump to:
Product Details & Documents
Alcohol-based rapid disinfectant wipes with comprehensive effectiveness for use in the Bacillol® wipes dispenser.
- Ready-to-use disinfectant wipes soaked in Bacillol® AF for use in the Bacillol® Wipes dispenser
- Free of dyes, perfumes and aldehydes
- ECARF-certified: professional allergy- and asthmatic-friendly surface disinfectants
- Can be used without gloves
- Fast and broadly effective
- Good wetting and fast drying
- Tear-resistant and absorbent PET fleece
- Wipes in dispenser can be used for 28 days
Can be used in the Bacillol® Wipes dispenser for quick disinfection of alcohol-resistant surfaces and medical inventory. In the healthcare sector, in laboratories as well as in commercial kitchens and in the food sector. In areas where a fast exposure and drying time is required.
Certificates
Search by Lot NumberSpecifications
- Colour:White
- Description:Stand-up pouches (90 wipes)
- Seq:02