ContactUsLinkComponent
Molecular sieve A5 (0.5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Molecular sieve A5 (0.5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Catalog # 20619.
Supplier:  Merck
CAS Number:  
Molecular sieve A5 (0.5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Catalog # 20619.
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
Specifications
About this item
 :
CAS Number:69912-79-4
 :1318-02-1