ContactUsLinkComponent
Cerex® OFXQ 96 Well Plate, RCH
Catalog # TCAT635-0101RCH
Supplier:  Tecan
Cerex® OFXQ 96 Well Plate, RCH
Catalog # TCAT635-0101RCH
Supplier:  Tecan
Specifications
  • Description:
    Plate RCH Cerex OFXQ
  • Environmentally Preferable:
  • Phase:
    OFXQ
  • Pk:
    1
Specifications
About this item
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.