Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid 1% (v/v) in aqueous solution
Acetic acid 1% (v/v) in aqueous solution
Acetic acid 1% (v/v) in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Acetic acid 1% (v/v) in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
64-19-7
:
Formula:
H₃CCOOH
MDL Number:
MFCD00036152
:
60.05 g/mol
Storage Temperature:
Ambient
EINECS:
200-580-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA035569
Videos
Thermo Scientific Acroseal 25 Years
Documentation