Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethanol
Ethanol ≥70% (v/v), extra pure
Ethanol ≥70% (v/v), extra pure
Flammable
Ethanol ≥70% (v/v), extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Flammable
Ethanol ≥70% (v/v), extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
64-17-5
:
Formula:
C₂H₅OH
MDL Number:
MFCD00003568
:
46.07 g/mol
Storage Temperature:
Ambient
EINECS:
200-578-6
UN:
1170
ADR:
3,III
:
GHS02