Order Entry
Germany
ContactUsLinkComponent
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Flammable
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier:  Merck
   Sustainable Options Available
Certificates
Flammable
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay (Ethanol):
    19.5 - 20.5 % (w/w)
  • Total aerobic microbial count (TAMC):
    ≤ 10 CFU/ml
  • Total combined yeasts/moulds count (TYMC):
    ≤ 10 CFU/ml
  • Bacterial endotoxins:
    ≤ 2.5 I.U./ml
  • Evaporation residue:
    ≤ 10 mg/l

Specification Test Results

Related Information

Frequently Bought Together