Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluorocytosine
5-Fluorocytosine
5-Fluorocytosine
MOLE33980289-1G
:
Molekula
:
5-Fluorocytosine
MOLE33980289-1G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
129.09 g/mol
:
13,04151
:
298 °C (1013 hPa)
:
165 °C
:
217-968-7
:
2022-85-7
:
C₄H₄FN₃O
:
MFCD00006035