Cytosine
Catalog # MOLEM40979102
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:111.1 g/mol
Merck Index:13,02824
Melting Pt:313…316 °C
Density:0.99771 g/cm³ (25 °C)
EINECS:200-749-5
:GHS07
Storage Temperature:Ambient
CAS Number:71-30-7
Formula:C₄H₅N₃O
MDL Number:MFCD00006034