3-Amino-4-methylcaproic acid
Marketsource
Supplier: BIONET RESEARCH
Jump to:
About this item
- CAS Number:40469-87-2
- Formula:C₇H₁₅NO₂
- MDL Number:MFCD03305808
- Molecular Weight:145.2 g/mol
4 Options Available Below
Product Details & Documents
- CAS Number:40469-87-2
- Formula:C₇H₁₅NO₂
- MDL Number:MFCD03305808
- Molecular Weight:145.2 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



