Order Entry
Germany
ContactUsLinkComponent
Accessories for High-Pressure Dosing Pump with 250 ml/min Stainless Steel Pump Head
Accessories for High-Pressure Dosing Pump with 250 ml/min Stainless Steel Pump Head
Catalog # KNAUAZZ00AB-1
Supplier:  KNAUER
Accessories for High-Pressure Dosing Pump with 250 ml/min Stainless Steel Pump Head
Catalog # KNAUAZZ00AB-1
Supplier:  KNAUER

Specifications

  • Description:
    Ternary LPG module for flow rates up to 220 ml/min (stainless steel)
  • Pk:
    1

Specifications

About this item