Order Entry
Germany
ContactUsLinkComponent
2-Octynoic acid methyl ester
2-Octynoic acid methyl ester
Catalog # EHERC15715700
Supplier:  EHRENSTORFER
CAS Number:  
2-Octynoic acid methyl ester
Catalog # EHERC15715700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :154.21 g/mol
EINECS:203-836-6
CAS Number:111-12-6
Formula:C₉H₁₄O₂
MDL Number:MFCD00009530