2-Octynoic acid methyl ester, Sigma-Aldrich®
Catalog # SIAL277509-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:154.21 g/mol
EINECS:203-836-6
CAS Number:111-12-6
Formula:C₉H₁₄O₂
MDL Number:MFCD00009530