2-Octynoic acid methyl ester ≥98%
Catalog # B21242.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
CAS Number:111-12-6
:
Formula:C₉H₁₄O₂
Boiling Pt:216…218 °C
Density:0.920
Flash Pt:88 °C (192 °F)
:154.21 g/mol
EINECS:203-836-6
:GHS06
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00009530