Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxynicotinic acid
2-Methoxynicotinic acid
2-Methoxynicotinic acid
2-Methoxynicotinic acid
Supplier:
Thermo Scientific
Certificates
Expand image
2-Methoxynicotinic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
16498-81-0
:
Formula:
C₇H₇NO₃
MDL Number:
MFCD00085110
:
153.14 g/mol