Ammonium iron (III) citrate (16.5 - 18.5% Fe)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1185-57-5
:
Formula:C₆H₅+4y)Fe(x)N(y)O₇
MDL Number:MFCD00013099
Storage Temperature:Ambient
EINECS:214-686-6
Specification Test Results
- Appearance:Brown-yellow to brown Powder
- Infrared spectrum:Conforms
- Assay:16.5 to 18.5 % (Fe)