Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Ethoxynicotinic acid

2-Ethoxynicotinic acid

Marketsource
Hersteller:   FLUOROCHEM
2-Ethoxynicotinic acid
Hersteller:  FLUOROCHEM
Marketsource
Certificates
2-Ethoxynicotinic acid

3 Options Available Below

Product Details & Documents
  • CAS Number:35969-54-1
  • Formula:C₈H₉NO₃
  • MDL Number:MFCD00128788
  • Molecular Weight:167.16 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Verfügbarkeit
    Preis
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 737,00 €
    737,00 €
    1 * 100 g
       737,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.