Anethole
Catalog # MOLEM52441337
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:148.2 g/mol
Merck Index:13,00647
Boiling Pt:234.5 °C (1013 hPa)
Melting Pt:21.3 °C
Density:1 g/cm³ (12 °C)
EINECS:203-205-5
CAS Number:104-46-1
Formula:C₁₀H₁₂O
MDL Number:MFCD00009284