Dytek® EP diamine (1,3-Diaminopentane) ≥98%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:589-37-7
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00134550
:102.18 g/mol
Boiling Pt:164 °C (lit.)
Melting Pt:–121 °C (lit.)
Density:0.855 g/mL (25 °C) (lit.)
UN:2734
ADR:8 (3),I
:M-Clarity™ quality level = MQ100
:GHS02
:GHS05