Order Entry
Germany
ContactUsLinkComponent
(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®
Irritant
(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
(S)-(-)-β-Citronellol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    156.27 g/mol
  • Boiling Point:
    109 - 111 °C (13 hPa)
  • Flash Point:
    101 °C DIN 51758
  • Refractive Index:
    1.4562 (20 °C, 589 nm)
  • Density:
    0.855 g/cm3 (20 °C)
  • Ignition Point:
    240 °C
  • Synonyms:
    (S)-(-)-3,7-Dimethyl-6-octen-1-ol, ß-Rhodinol
  • Hill Formula:
    C10H20O

Specification Test Results

Related Information