FreSR™-S Single Cell Freezing Medium
Catalog # STMC05859
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:50 mL
- Description:FreSR™-S, 50 ml
- Format:Human; animal component-free, serum-free
Specifications
About this item
Animal component-free medium for freezing ES and iPS cells as single cells.
- Defined, serum-free and animal component-free medium for cryopreserving ES/iPS cells as single cells
- Quickly recover ES/iPS cell colonies after thawing
- Reproducibly high recovery rates
- Optimised for cryopreserving ES/iPS cells cultured in TeSR™ maintenance media
- Preserves ES/iPS cell pluripotency and expansion capacities
- Convenient, ready-to-use format
FreSR™-S is a defined, serum-free, and animal component-free medium for the cryopreservation of human pluripotent stem cells (hPSCs) as single cells. This complete and ready-to-use medium is recommended for hPSCs cultured in mTeSR™1, mTeSR™ Plus, TeSR™2, or TeSR™-E8™. Cryopreserved hPSCs should be stored at −135 °C (liquid nitrogen) or colder.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').