ContactUsLinkComponent
Communicating tubes
Communicating tubes
Catalog # 765-0969
undefined
Communicating tubes
Catalog # 765-0969
Specifications
  • Description:
    Communicating tube with foot
  • Pk:
    1
Specifications
About this item
Five glass tubes of different shape and inner dimensions. Down to each other. When filling the liquid in the pipes, it is clear that the height of the stowage in the tubes. pipes are independent of the design of the pipes. Comes with foot.