Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Chlorophenyl)propionic acid

3-(2-Chlorophenyl)propionic acid

Marketsource
Supplier:   Chemicals
3-(2-Chlorophenyl)propionic acid
Supplier:  Chemicals
Marketsource
Certificates
3-(2-Chlorophenyl)propionic acid

3 Options Available Below

Product Details & Documents
Synonyms:  3-(2-Chlorophenyl)propanoic acid
  • CAS Number:1643-28-3
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00016547
  • Molecular Weight:184.62 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

This product has been blocked by your organization. Please contact your purchasing department for more information