Order Entry
Germany
Orders LinkContactUsLinkComponent
Water

Water

Glass bottle, 2 mL
Catalog # TREV4850-20-05
Marketsource
Supplier:   TREVIGEN
Water
Catalog # TREV4850-20-05
Supplier:  TREVIGEN
Marketsource
CAS Number:  7732-18-5
Water

Glass bottle, 2 mL

CAS Number:  7732-18-5
Price
Inquire For Price
Log in for availability
1 * 2 mL
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL

Specifications