Order Entry
Germany
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Irritant
Marketsource
Supplier:   TREVIGEN
Propidium Iodide
Supplier:  TREVIGEN
Marketsource
Certificates
Propidium Iodide

2 Options Available Below

Product Details & Documents
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • EINECS:247-081-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 mL
    Log in for availability
    Log in for availability
    1 * 2,5 mL
    1 * 2,5 mL

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.