Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vinyltrimethylsilane
Vinyltrimethylsilane
Vinyltrimethylsilane
Vinyltrimethylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Vinyltrimethylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
754-05-2
:
Formula:
C₅H₁₂Si
MDL Number:
MFCD00008606
Boiling Pt:
55 °C (1013 hPa)
Density:
0.7038 g/cm³ (20 °C)
:
100.24 g/mol
EINECS:
212-042-9