Vinyltrimethylsilane
Catalog # CHMPSILS20480.
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
Boiling Pt:55 °C (1013 hPa)
Density:0.7038 g/cm³ (20 °C)
:100.24 g/mol
EINECS:212-042-9
CAS Number:754-05-2
Formula:C₅H₁₂Si
MDL Number:MFCD00008606