Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trimethylsilyl)-1-propanol
3-(Trimethylsilyl)-1-propanol
3-(Trimethylsilyl)-1-propanol
3-(Trimethylsilyl)-1-propanol
Supplier:
CHEMPUR SILANE
Certificates
Expand image
3-(Trimethylsilyl)-1-propanol
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
2917-47-7
:
Formula:
HOH₂CCH₂CH₂Si(CH₃)₃
:
132.28 g/mol
Boiling Pt:
111 °C (1013 hPa)
Density:
0.8324 g/cm³ (20 °C)