Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ethoxytrimethylsilane

Marketsource
Supplier:   CHEMPUR SILANE
Ethoxytrimethylsilane
Supplier:  CHEMPUR SILANE
Marketsource
Certificates
Ethoxytrimethylsilane

2 Options Available Below

Product Details & Documents
Synonyms:  Trimethyl(ethoxy)silane
  • CAS Number:1825-62-3
  • Formula:C₅H₁₄OSi
  • MDL Number:MFCD00009069
  • Molecular Weight:118.25 g/mol
  • Boiling Pt:75 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0,7573 g/cm³ (20 °C)
  • EINECS:217-370-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource